Gene Transcripts

Gene Symbol
Ugt2b15
Entrez Gene ID
83808
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide B15
Gene Aliases
Ugt2b12, Ugt2b36, Ugt2b4
Chromosome Location
Chr. 14: 22553635 - 22591461 on Build Rnor_6.0
UniGene ID
Rn.3686
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004271.1 NP_001004271.1 1 240
XM_008770060.1 XP_008768282.1 1 267
GenBank mRNA BC078782.1 - 1 240
U06273.1 - 1 221

View Full Product Details