Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102431.1 | NP_001095901.1 | 1 | 208 | |
XM_006235056.2 | XP_006235118.1 | 2 | 364 | ||
XM_006235057.2 | XP_006235119.1 | 2 | 364 | ||
XM_006235058.2 | XP_006235120.1 | 2 | 318 | ||
GenBank mRNA | BC079436.1 | - | 1 | 220 | |
GQ245669.1 | - | 1 | 208 |