Gene Transcripts

Gene Symbol
Exoc4
Entrez Gene ID
116654
Gene Name
exocyst complex component 4
Gene Aliases
ALR, Sec8, Sec8l1
Chromosome Location
Chr. 4: 60549129 - 61358305 on Build Rnor_6.0
UniGene ID
Rn.51841
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053875.1 NP_446327.1 2 257
XM_008762757.1 XP_008760979.1 2 325
GenBank mRNA U32498.1 - 2 257

View Full Product Details