Gene Transcripts

Gene Symbol
Paics
Entrez Gene ID
140946
Gene Name
phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase
Gene Aliases
Ade2h1, Airc
Chromosome Location
Chr. 14: 33563884 - 33580924 on Build Rnor_6.0
UniGene ID
Rn.3015
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080910.3 NP_543186.1 6, 7 790
XM_006250852.2 XP_006250914.1 6, 7 838
XM_006250853.2 XP_006250915.1 6, 7 809
XM_006250854.2 XP_006250916.1 6, 7 839
GenBank mRNA BC072508.1 - 6, 7 780
BC085711.1 - 6, 7 790 7192781
D37979.1 - 4, 5 695

Gene Symbol
LOC100910308
Entrez Gene ID
100910308
Gene Name
multifunctional protein ADE2-like
Gene Aliases
Chromosome Location
Chr. 14: 33968258 - 33977352 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751352.3 XP_003751400.2 3, 4 566

View Full Product Details