Gene Transcripts

Gene Symbol
Bles03
Entrez Gene ID
266609
Gene Name
basophilic leukemia expressed protein BLES03
Gene Aliases
Chromosome Location
Chr. 1: 220806563 - 220808817 on Build Rnor_6.0
UniGene ID
Rn.7405
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024233.2 NP_001019404.2 2 416
NM_001243534.1 NP_001230463.1 2 430
GenBank mRNA BC093387.1 - 2 275
CK483724.1 - 2 272
CK596744.1 - 2 416
FQ233132.1 - 2 309

View Full Product Details