Gene Transcripts

Gene Symbol
SEC62
Entrez Gene ID
7095
Gene Name
SEC62 homolog, preprotein translocation factor
Gene Aliases
Dtrp1, HTP1, TLOC1, TP-1
Chromosome Location
Chr. 3: 169966792 - 169998373 on Build GRCh38
UniGene ID
Hs.744859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003262.3 NP_003253.1 2 124
XM_011513114.2 XP_011511416.1 3 201
GenBank mRNA BC003677.1 - 2 116
BC007040.1 - 2 116
BC012035.1 - 2 108 4706586
BC105971.1 - 207
BX475495.1 - 2 125
BX537668.1 - 1 8087
BX648539.1 - 2 108
D87127.1 - 2 105
KC005990.1 - 2 93
U93239.1 - 2 105

View Full Product Details