Gene Transcripts

Gene Symbol
Rilp
Entrez Gene ID
287531
Gene Name
Rab interacting lysosomal protein
Gene Aliases
Chromosome Location
Chr. 10: 63659030 - 63662188 on Build Rnor_6.0
UniGene ID
Rn.84076
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105811.1 NP_001099281.1 7 963

View Full Product Details