Gene Transcripts

Gene Symbol
Ankrd45
Entrez Gene ID
289152
Gene Name
ankyrin repeat domain 45
Gene Aliases
RGD1311153
Chromosome Location
Chr. 13: 78973424 - 79000332 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006250119.2 XP_006250181.1 3 672
XM_006250122.2 XP_006250184.1 3 667
XM_006250123.2 XP_006250185.1 3 663
XM_008769703.1 XP_008767925.1 5 728
XM_008769704.1 XP_008767926.1 3 664

View Full Product Details