Gene Transcripts

Gene Symbol
Mptx
Entrez Gene ID
289243
Gene Name
mucosal pentraxin
Gene Aliases
Mptx1
Chromosome Location
Chr. 13: 91773004 - 91776397 on Build Rnor_6.0
UniGene ID
Rn.160486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037642.1 NP_001032731.1 2 153
GenBank mRNA AY426671.1 - 2 153

View Full Product Details