Gene Transcripts
- Gene Symbol
- Sowahb
- Entrez Gene ID
- 289501
- Gene Name
- sosondowah ankyrin repeat domain family member B
- Gene Aliases
- Ankrd56, RGD1311994
- Chromosome Location
- Chr. 14: 16491023 - 16495764 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_223228.4 |
XP_223228.1
|
1 |
2458 |
|
View Full Product Details