Gene Transcripts

Gene Symbol
St8sia6
Entrez Gene ID
291325
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Gene Aliases
Siat8f
Chromosome Location
Chr. 17: 80960785 - 81002830 on Build Rnor_6.0
UniGene ID
Rn.104457
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213624.1 NP_998789.1 816
GenBank mRNA AJ699423.1 - 816

View Full Product Details