Gene Transcripts

Gene Symbol
UBE2D1
Entrez Gene ID
7321
Gene Name
ubiquitin conjugating enzyme E2 D1
Gene Aliases
E2(17)KB1, SFT, UBC4/5, UBCH5, UBCH5A
Chromosome Location
Chr. 10: 58334979 - 58370753 on Build GRCh38
UniGene ID
Hs.129683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204880.1 NP_001191809.1 3 355
NM_003338.4 NP_003329.1 4 387
XM_017016609.1 XP_016872098.1 4 286
GenBank mRNA AF257220.1 - 4 310
AJ272367.1 - 4 310
AK291901.1 - 4 358
BC005980.1 - 4 358 4106710
BC015997.1 - 4 362 4305900
BI598342.1 - 4 391
BT007041.1 - 4 164
DA618122.1 - 3 328
DA677507.1 - 4 387
X78140.1 - 4 179

View Full Product Details