Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100517.1 | NP_001093987.1 | 16 | 2304 | |
XM_006255580.2 | XP_006255642.1 | 16 | 2542 | ||
XM_006255581.2 | XP_006255643.1 | 17 | 2399 | ||
XM_006255582.2 | XP_006255644.1 | 16 | 2538 | ||
XM_006255583.2 | XP_006255645.1 | 16 | 2294 | ||
GenBank mRNA | BC098738.1 | - | 3 | 416 |