Gene Transcripts

Gene Symbol
UPK3A
Entrez Gene ID
7380
Gene Name
uroplakin 3A
Gene Aliases
UP3A, UPIII, UPIIIA, UPK3
Chromosome Location
Chr. 22: 45284987 - 45295874 on Build GRCh38
UniGene ID
Hs.632787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006953.3 NP_008884.1 3, 4 515
GenBank mRNA AB010116.1 - 2, 3 392
AB010637.1 - 3, 4 488
AF085808.1 - 3, 4 520
BC069544.1 - 3, 4 520 7262218
CR456608.1 - 3, 4 490

View Full Product Details