Gene Transcripts

Gene Symbol
Ankrd22
Entrez Gene ID
294093
Gene Name
ankyrin repeat domain 22
Gene Aliases
Chromosome Location
Chr. 1: 252430750 - 252461346 on Build Rnor_6.0
UniGene ID
Rn.220761
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191638.1 NP_001178567.1 5, 6 641

View Full Product Details