Gene Transcripts

Gene Symbol
Trim31
Entrez Gene ID
294208
Gene Name
tripartite motif-containing 31
Gene Aliases
Trim31l
Chromosome Location
Chr. 20: 2146245 - 2159875 on Build Rnor_6.0
UniGene ID
Rn.124521
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106376.1 NP_001099846.1 1 347
XM_008772691.1 XP_008770913.1 2 258

View Full Product Details