Gene Transcripts

Gene Symbol
Pi16
Entrez Gene ID
294312
Gene Name
peptidase inhibitor 16
Gene Aliases
Chromosome Location
Chr. 20: 6923173 - 6932786 on Build Rnor_6.0
UniGene ID
Rn.9448
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170481.1 NP_001163952.1 2 364
GenBank mRNA BC100265.1 - 2 364
FQ220139.1 - 2 373

View Full Product Details