Gene Transcripts

Gene Symbol
Tm4sf1
Entrez Gene ID
295061
Gene Name
transmembrane 4 L six family member 1
Gene Aliases
Chromosome Location
Chr. 2: 147382866 - 147392189 on Build Rnor_6.0
UniGene ID
Rn.24712
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106434.1 NP_001099904.1 1 403
XM_008760989.1 XP_008759211.1 1 530
GenBank mRNA FQ220762.1 - 1 169

View Full Product Details