Gene Transcripts

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
Chromosome Location
Chr. 10: 109864766 - 109923553 on Build GRCh38
UniGene ID
Hs.390623
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324128.1 NP_001311057.1 13 1326
NM_001324131.1 NP_001311060.1 15 1523
NM_001324132.1 NP_001311061.1 14 1390
NM_001324133.1 NP_001311062.1 15 1479
NM_001324134.1 NP_001311063.1 13 1310
NM_001324135.1 NP_001311064.1 14 1428
NM_001324136.1 NP_001311065.1 15 1694
NM_020383.3 NP_065116.3 15 1479
XM_011540134.1 XP_011538436.1 15 1473
XM_017016613.1 XP_016872102.1 14 1329
XM_017016614.1 XP_016872103.1 16 1462
XM_017016615.1 XP_016872104.1 14 1346
GenBank mRNA AF195530.1 - 14 1306
AF272981.1 - 13 1230
AK093693.1 - 13 1284
AK095447.1 - 1 1699
AK223513.1 - 14 1290
AK289436.1 - 15 1382
BC005126.2 - 14 1268 3836436
BC007579.1 - 15 1370 3139868
BC013417.2 - 11 980
BX537682.1 - 11 3324
CR456922.1 - 13 1230
X95762.1 - 13 1230

View Full Product Details