Gene Transcripts

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
Chromosome Location
Chr. 10: 109864766 - 109923553 on Build GRCh38
UniGene ID
Hs.390623
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167604.1 NP_001161076.1 3 314
NM_001324128.1 NP_001311057.1 2 225
NM_001324131.1 NP_001311060.1 3 314
NM_001324132.1 NP_001311061.1 2 225
NM_001324133.1 NP_001311062.1 3 314
NM_001324134.1 NP_001311063.1 3 314
NM_001324135.1 NP_001311064.1 3 314
NM_001324136.1 NP_001311065.1 3 529
NM_020383.3 NP_065116.3 3 314
XM_011540134.1 XP_011538436.1 3 308
XM_017016613.1 XP_016872102.1 3 308
XM_017016614.1 XP_016872103.1 3 202
XM_017016615.1 XP_016872104.1 3 245
GenBank mRNA AF195530.1 - 2 141
AF272981.1 - 1 65
AK093693.1 - 2 183
AK223513.1 - 2 125
AK289436.1 - 3 217
AK304367.1 - 3 207
AL833411.1 - 3 220
BC005126.2 - 2 103 3836436
BC007579.1 - 3 205 3139868
BX537682.1 - 2 139
CR456922.1 - 1 65
DA484161.1 - 2 221
X95762.1 - 1 65

View Full Product Details