Gene Transcripts

Gene Symbol
Sema3e
Entrez Gene ID
296789
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E
Gene Aliases
Chromosome Location
Chr. 4: 17314745 - 17594598 on Build Rnor_6.0
UniGene ID
Rn.214084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106579.1 NP_001100049.1 3 910
XM_008762698.1 XP_008760920.1 2 678

View Full Product Details