Gene Transcripts

Gene Symbol
Ccdc37
Entrez Gene ID
297444
Gene Name
coiled-coil domain containing 37
Gene Aliases
RGD1305768
Chromosome Location
Chr. 4: 122314475 - 122339007 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006236885.2 XP_006236947.1 14 2254
XM_006236886.2 XP_006236948.1 13 2078

View Full Product Details