Gene Transcripts

Gene Symbol
Trabd
Entrez Gene ID
300142
Gene Name
TraB domain containing
Gene Aliases
RGD1310875
Chromosome Location
Chr. 7: 130058252 - 130069224 on Build Rnor_6.0
UniGene ID
Rn.3565
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106788.1 NP_001100258.1 4 372
XM_006242182.2 XP_006242244.1 4 934
XM_006242183.2 XP_006242245.1 4 787

View Full Product Details