Gene Transcripts

Gene Symbol
Srprb
Entrez Gene ID
300965
Gene Name
signal recognition particle receptor, B subunit
Gene Aliases
Ab2-417, Ba1-667, Cc1-8
Chromosome Location
Chr. 8: 111673479 - 111687426 on Build Rnor_6.0
UniGene ID
Rn.138078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013252.1 NP_001013270.1 6 590
GenBank mRNA AY325214.1 - 2738
AY325230.1 - 2741
AY325241.1 - 2738
BC098951.1 - 582 7324816
BF523222.1 - 3 256

View Full Product Details