Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_057176.2 | NP_476517.1 | 4 | 1292 | |
GenBank mRNA | AK129999.1 | - | 4 | 1303 | |
BC103898.1 | - | 4 | 1271 | 40005137 | |
BC103899.1 | - | 4 | 1271 | 40005138 | |
BC103900.1 | - | 4 | 1271 | 40005139 |