Gene Transcripts

Gene Symbol
Cnnm3
Entrez Gene ID
301345
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Gene Aliases
Chromosome Location
Chr. 9: 43093128 - 43108486 on Build Rnor_6.0
UniGene ID
Rn.50710
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106901.2 NP_001100371.2 4 1718
XM_006244756.2 XP_006244818.1 4 1725
XM_006244757.2 XP_006244819.1 4 1728

View Full Product Details