Gene Transcripts

Gene Symbol
ALDH5A1
Entrez Gene ID
7915
Gene Name
aldehyde dehydrogenase 5 family member A1
Gene Aliases
SSADH, SSDH
Chromosome Location
Chr. 6: 24494969 - 24537207 on Build GRCh38
UniGene ID
Hs.371723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080.3 NP_001071.1 2 414
NM_170740.1 NP_733936.1 2 414
GenBank mRNA AJ427353.1 - 2 182
AJ427354.2 - 2 182
AJ427355.2 - 2 386
AK290800.1 - 2 532
AK294699.1 - 3 191
AK314357.1 - 2 414
AK315380.1 - 2 509
BC034321.1 - 2 414 4829533
KJ534766.1 - 2 386
KJ534767.1 - 2 386
KJ534768.1 - 2 386
KJ534769.1 - 2 386
Y11192.1 - 2 386

View Full Product Details