Gene Transcripts

Gene Symbol
Sept11
Entrez Gene ID
305227
Gene Name
septin 11
Gene Aliases
Sept6
Chromosome Location
Chr. 14: 16369544 - 16405645 on Build Rnor_6.0
UniGene ID
Rn.27798
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107208.2 NP_001100678.1 45
GenBank mRNA BC167769.1 - 176
EU711414.1 - 45
EU711415.1 - 45
EU711416.1 - 45
EU711417.1 - 45

View Full Product Details