Gene Transcripts

Gene Symbol
Ptpn20b
Entrez Gene ID
306281
Gene Name
protein tyrosine phosphatase, non-receptor type 20B
Gene Aliases
Ptpn20
Chromosome Location
Chr. 16: 9907607 - 9973514 on Build Rnor_6.0
UniGene ID
Rn.128630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080149.1 NP_001073618.1 9 1149
XM_006252765.2 XP_006252827.1 10 1222
XM_006252767.2 XP_006252829.1 10 1189
XM_008771099.1 XP_008769321.1 10 1175
GenBank mRNA BC129117.1 - 9 1149

View Full Product Details