Gene Transcripts

Gene Symbol
Spg11
Entrez Gene ID
311372
Gene Name
spastic paraplegia 11 (autosomal recessive)
Gene Aliases
RGD1562529
Chromosome Location
Chr. 3: 113999580 - 114064744 on Build Rnor_6.0
UniGene ID
Rn.228765
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191571.1 NP_001178500.1 33 6118
XM_006234855.2 XP_006234917.1 35 6812

View Full Product Details