Gene Transcripts

Gene Symbol
Rbm28
Entrez Gene ID
312182
Gene Name
RNA binding motif protein 28
Gene Aliases
Chromosome Location
Chr. 4: 56398769 - 56438446 on Build Rnor_6.0
UniGene ID
Rn.50022
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107850.1 NP_001101320.1 14 1541
XM_006236190.2 XP_006236252.1 16 1876
GenBank mRNA FQ211470.1 - 15 1641

View Full Product Details