Gene Transcripts

Gene Symbol
Ano6
Entrez Gene ID
315272
Gene Name
anoctamin 6
Gene Aliases
Tmem16f
Chromosome Location
Chr. 7: 137142067 - 137335208 on Build Rnor_6.0
UniGene ID
Rn.3350
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108108.1 NP_001101578.1 8, 9 1013
XM_008765735.1 XP_008763957.1 8, 9 1116
XM_008765736.1 XP_008763958.1 7, 8 1053

View Full Product Details