Gene Transcripts

Gene Symbol
Slc22a14
Entrez Gene ID
316061
Gene Name
solute carrier family 22, member 14
Gene Aliases
Chromosome Location
Chr. 8: 127858425 - 127871192 on Build Rnor_6.0
UniGene ID
Rn.129260
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108193.1 NP_001101663.1 3 698
GenBank mRNA BC168201.1 - 801

View Full Product Details