Gene Transcripts
- Gene Symbol
- Sap30l
- Entrez Gene ID
- 360531
- Gene Name
- SAP30-like
- Gene Aliases
- RGD1309585
- Chromosome Location
- Chr. 10: 43224151 - 43231864 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_340803.7 |
XP_340804.4
|
4 |
566 |
|
View Full Product Details