Gene Transcripts

Gene Symbol
Unk
Entrez Gene ID
360663
Gene Name
unkempt family zinc finger
Gene Aliases
Zc3h5, Zc3hdc5
Chromosome Location
Chr. 10: 104582531 - 104613102 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750953.3 XP_003751001.1 3 852
XM_003750954.3 XP_003751002.1 3 455
XM_006247762.2 XP_006247824.1 3 455

View Full Product Details