Gene Transcripts

Gene Symbol
ASGR1
Entrez Gene ID
432
Gene Name
asialoglycoprotein receptor 1
Gene Aliases
ASGPR, ASGPR1, CLEC4H1, HL-1
Chromosome Location
Chr. 17: 7173431 - 7179564 on Build GRCh38
UniGene ID
Hs.12056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197216.2 NP_001184145.1 8 1253
NM_001671.4 NP_001662.1 9 1370
XM_011523861.2 XP_011522163.1 9 1277
GenBank mRNA AB070933.1 - 1149
AK308375.1 - 9 1174
BC032130.2 - 9 1047 5087444
BM818592.1 - 1 170
M10058.1 - 9 1141

View Full Product Details