Gene Transcripts

Gene Symbol
Riok3
Entrez Gene ID
361293
Gene Name
RIO kinase 3
Gene Aliases
Chromosome Location
Chr. 18: 3565166 - 3590739 on Build Rnor_6.0
UniGene ID
Rn.203135
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108423.1 NP_001101893.1 9 2041
GenBank mRNA FQ230531.1 - 6 817

View Full Product Details