Gene Transcripts

Gene Symbol
Sh3yl1
Entrez Gene ID
362724
Gene Name
SH3 and SYLF domain containing 1
Gene Aliases
Chromosome Location
Chr. 6: 49851742 - 49895502 on Build Rnor_6.0
UniGene ID
Rn.1163
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108705.2 NP_001102175.1 10 1074
XM_006239970.2 XP_006240032.1 10 1536
GenBank mRNA BC158742.1 - 10 1074

View Full Product Details