Gene Transcripts
- Gene Symbol
- Bex2
- Entrez Gene ID
- 363498
- Gene Name
- brain expressed X-linked 2
- Gene Aliases
- EG2RVC
- Chromosome Location
- Chr. X: 106556838 - 106558366 on Build Rnor_6.0
- UniGene ID
- Rn.33240
- Species
- Rattus norvegicus
- Gene Symbol
- Bex1
- Entrez Gene ID
- 501625
- Gene Name
- brain expressed, X-linked 1
- Gene Aliases
- Chromosome Location
- Chr. X: 106761388 - 106771656 on Build Rnor_6.0
- UniGene ID
- Rn.9190
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY208292.1 |
- |
|
641 |
|
View Full Product Details