Gene Transcripts

Gene Symbol
Bex2
Entrez Gene ID
363498
Gene Name
brain expressed X-linked 2
Gene Aliases
EG2RVC
Chromosome Location
Chr. X: 106556838 - 106558366 on Build Rnor_6.0
UniGene ID
Rn.33240
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077435.1 NP_001070903.1 3 286
GenBank mRNA AY833555.1 - 3 286

View Full Product Details