Gene Transcripts

Gene Symbol
Ap1m2
Entrez Gene ID
367038
Gene Name
adaptor-related protein complex 1, mu 2 subunit
Gene Aliases
RGD1561490
Chromosome Location
Chr. 8: 22318941 - 22336827 on Build Rnor_6.0
UniGene ID
Rn.216511
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108996.2 NP_001102466.1 9 971
GenBank mRNA BC167082.1 - 9 971

View Full Product Details