Gene Transcripts

Gene Symbol
Ly49s4
Entrez Gene ID
494195
Gene Name
Ly49 stimulatory receptor 4
Gene Aliases
Chromosome Location
Chr. 4: 164470396 - 164493252 on Build Rnor_6.0
UniGene ID
Rn.198707
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009487.1 NP_001009487.1 432
GenBank mRNA AY649831.1 - 432

View Full Product Details