Gene Transcripts

Gene Symbol
Vom1r66
Entrez Gene ID
494291
Gene Name
vomeronasal 1 receptor 66
Gene Aliases
V1rc19
Chromosome Location
Chr. 4: 87516218 - 87517162 on Build Rnor_6.0
UniGene ID
Rn.129674
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008945.1 NP_001008945.1 1 421
GenBank mRNA AY510310.1 - 1 421

View Full Product Details