Gene Transcripts

Gene Symbol
Rab11fip1
Entrez Gene ID
498650
Gene Name
RAB11 family interacting protein 1 (class I)
Gene Aliases
RGD1562965, Rab11-FIP1
Chromosome Location
Chr. 16: 69048739 - 69079332 on Build Rnor_6.0
UniGene ID
Rn.49288
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191555.1 NP_001178484.1 4 1866
XM_008771331.1 XP_008769553.1 4 2123

View Full Product Details