Gene Transcripts

Gene Symbol
Armc2
Entrez Gene ID
499470
Gene Name
armadillo repeat containing 2
Gene Aliases
RGD1562413
Chromosome Location
Chr. 20: 46766467 - 46872010 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006256579.2 XP_006256641.1 12, 13 1805
XM_574794.4 XP_574794.3 12, 13 1718

View Full Product Details