Gene Transcripts

Gene Symbol
Bpifb3
Entrez Gene ID
499924
Gene Name
BPI fold containing family B, member 3
Gene Aliases
Lplunc3, Rya3
Chromosome Location
Chr. 3: 149401890 - 149416506 on Build Rnor_6.0
UniGene ID
Rn.218676
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109208.1 NP_001102678.1 8 822
XM_006235352.2 XP_006235414.1 7 724
GenBank mRNA X60658.1 - 8 788

View Full Product Details