Gene Transcripts

Gene Symbol
Bmp8b
Entrez Gene ID
679119
Gene Name
bone morphogenetic protein 8b
Gene Aliases
Chromosome Location
Chr. 5: 140803510 - 140821104 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729526.4 XP_002729572.1 7 1268

View Full Product Details