Gene Transcripts

Gene Symbol
Nkpd1
Entrez Gene ID
680440
Gene Name
NTPase, KAP family P-loop domain containing 1
Gene Aliases
Chromosome Location
Chr. 1: 80424507 - 80434115 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001057808.5 XP_001057808.2 5 3563
XM_006228500.2 XP_006228562.1 4 3116

View Full Product Details