Gene Transcripts

Gene Symbol
Loxhd1
Entrez Gene ID
291427
Gene Name
lipoxygenase homology domains 1
Gene Aliases
Chromosome Location
Chr. 18: 73645907 - 73812271 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772102.1 XP_008770324.1 39 5953

View Full Product Details