Gene Transcripts

Gene Symbol
Dennd1c
Entrez Gene ID
363326
Gene Name
DENN/MADD domain containing 1C
Gene Aliases
Chromosome Location
Chr. 9: 9976015 - 9982349 on Build Rnor_6.0
UniGene ID
Rn.162357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126289.1 NP_001119761.1 7, 8 919
GenBank mRNA BC160823.1 - 7, 8 919

View Full Product Details